siRNA Details
| Gene Symbol: Wisp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170483.1 | NP_001163954.1 | 3 | 383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wisp3| Gene Symbol: Wisp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170483.1 | NP_001163954.1 | 3 | 383 | ||