siRNA Details
| Gene Symbol: Ostm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029925.1 | NP_001025096.1 | 986 | |||
| XM_006256577.2 | XP_006256639.1 | 5, 6 | 1028 | |||
| GenBank mRNA | BC097978.1 | - | 986 | 7442765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ostm1| Gene Symbol: Ostm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029925.1 | NP_001025096.1 | 986 | |||
| XM_006256577.2 | XP_006256639.1 | 5, 6 | 1028 | |||
| GenBank mRNA | BC097978.1 | - | 986 | 7442765 | ||