siRNA Details
| Gene Symbol: Fam151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134748.1 | NP_001128220.1 | 1, 2 | 120 | ||
| GenBank mRNA | BC167070.1 | - | 1, 2 | 120 | 9032304 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam151b| Gene Symbol: Fam151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134748.1 | NP_001128220.1 | 1, 2 | 120 | ||
| GenBank mRNA | BC167070.1 | - | 1, 2 | 120 | 9032304 | |