siRNA Details
| Gene Symbol: Ccdc125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134761.1 | NP_001128233.1 | 5, 6 | 617 | ||
| GenBank mRNA | BC166734.1 | - | 1, 2 | 476 | 7936477 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc125| Gene Symbol: Ccdc125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134761.1 | NP_001128233.1 | 5, 6 | 617 | ||
| GenBank mRNA | BC166734.1 | - | 1, 2 | 476 | 7936477 | |