siRNA Details
| Gene Symbol: Ccno | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109175.1 | NP_001102645.1 | 1 | 1063 | ||
| XM_006231947.2 | XP_006232009.1 | 2 | 2195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccno| Gene Symbol: Ccno | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109175.1 | NP_001102645.1 | 1 | 1063 | ||
| XM_006231947.2 | XP_006232009.1 | 2 | 2195 | |||