siRNA Details
| Gene Symbol: LOC499530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047941.1 | NP_001041406.1 | 2 | 198 | ||
| XM_008760739.1 | XP_008758961.1 | 2 | 198 | |||
| GenBank mRNA | AY539940.1 | - | 2 | 198 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499530| Gene Symbol: LOC499530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047941.1 | NP_001041406.1 | 2 | 198 | ||
| XM_008760739.1 | XP_008758961.1 | 2 | 198 | |||
| GenBank mRNA | AY539940.1 | - | 2 | 198 | ||