siRNA Details
| Gene Symbol: Ccdc152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191959.1 | NP_001178888.1 | 7 | 599 | ||
| XM_006231988.2 | XP_006232050.1 | 8 | 677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc152| Gene Symbol: Ccdc152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191959.1 | NP_001178888.1 | 7 | 599 | ||
| XM_006231988.2 | XP_006232050.1 | 8 | 677 | |||