siRNA Details
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109176.1 | NP_001102646.1 | 14, 15 | 2321 | ||
| XM_006232019.2 | XP_006232081.1 | 15, 16 | 2601 | |||
| XM_008760784.1 | XP_008759006.1 | 15, 16 | 2602 | |||
| XM_008760785.1 | XP_008759007.1 | 15, 16 | 2591 | |||
| XM_008760786.1 | XP_008759008.1 | 16, 17 | 2695 | |||
| XM_008760787.1 | XP_008759009.1 | 15, 16 | 4055 | |||
| XM_008760788.1 | XP_008759010.1 | 15, 16 | 4150 | |||
| XM_008760789.1 | XP_008759011.1 | 14, 15 | 2463 | |||

Fyb
Genomic Map