siRNA Details
| Gene Symbol: Lmbrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109177.1 | NP_001102647.1 | 9 | 1345 | ||
| XM_006232024.2 | XP_006232086.1 | 9 | 1336 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_177178, BC125421, AK153728, AK036527, BC120727, and others.

Lmbrd2
Genomic Map