siRNA Details
Gene Symbol: LOC499541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047942.1 | NP_001041407.1 | 3 | 325 | ||
XM_008760791.1 | XP_008759013.1 | 6 | 1253 | |||
GenBank mRNA | AY383700.1 | - | 3 | 325 |
