siRNA Details
| Gene Symbol: Zfp458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024299.1 | NP_001019470.1 | 4 | 2097 | ||
| NM_001142962.1 | NP_001136434.1 | 4 | 1998 | |||
| GenBank mRNA | BC085784.1 | - | 4 | 2097 | 7107819 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp458| Gene Symbol: Zfp458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024299.1 | NP_001019470.1 | 4 | 2097 | ||
| NM_001142962.1 | NP_001136434.1 | 4 | 1998 | |||
| GenBank mRNA | BC085784.1 | - | 4 | 2097 | 7107819 | |