siRNA Details
| Gene Symbol: Zfp458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024299.1 | NP_001019470.1 | 4 | 1179 | ||
| NM_001142962.1 | NP_001136434.1 | 4 | 1080 | |||
| XM_008760843.1 | XP_008759065.1 | 4 | 1749 | |||
| GenBank mRNA | BC085784.1 | - | 4 | 1179 | 7107819 | |
| Gene Symbol: LOC102551340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227824.2 | XP_006227886.2 | 4 | 639 | ||
| XM_008758708.1 | XP_008756930.1 | 4 | 640 | |||

Zfp458
Genomic Map