siRNA Details
| Gene Symbol: Gpr160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025147.1 | NP_001020318.1 | 3 | 408 | ||
| XM_008760935.1 | XP_008759157.1 | 3 | 646 | |||
| XM_008760936.1 | XP_008759158.1 | 2 | 994 | |||
| GenBank mRNA | BC082060.1 | - | 3 | 408 | 7132467 | |

Gpr160
Genomic Map