siRNA Details
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044696.1 | NP_001038161.1 | 5 | 607 | ||
XM_006232235.2 | XP_006232297.1 | 6 | 578 | |||
XM_008760937.1 | XP_008759159.1 | 6 | 669 | |||
XM_008760938.1 | XP_008759160.1 | 5 | 365 | |||
XM_008760939.1 | XP_008759161.1 | 5 | 561 | |||
GenBank mRNA | BC097432.1 | - | 5 | 607 | 7455342 |
