siRNA Details
| Gene Symbol: Lrrc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044696.1 | NP_001038161.1 | 5 | 635 | ||
| XM_006232235.2 | XP_006232297.1 | 6 | 606 | |||
| XM_008760937.1 | XP_008759159.1 | 6 | 697 | |||
| XM_008760938.1 | XP_008759160.1 | 5 | 393 | |||
| XM_008760939.1 | XP_008759161.1 | 5 | 589 | |||
| GenBank mRNA | BC097432.1 | - | 5 | 635 | 7455342 | |

Lrrc34
Genomic Map