siRNA Details
| Gene Symbol: LOC499602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025039.2 | NP_001020210.2 | 11 | 1161 | ||
| XM_006232310.2 | XP_006232372.1 | 10 | 1291 | |||
| XM_006232311.2 | XP_006232373.1 | 7 | 828 | |||
| XM_008760940.1 | XP_008759162.1 | 10 | 1020 | |||
| GenBank mRNA | BC086604.1 | - | 1 | 303 | 7317121 | |
| BC098046.1 | - | 7 | 800 | |||

LOC499602
Genomic Map