siRNA Details
| Gene Symbol: LOC499607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760983.1 | XP_008759205.1 | 8 | 1123 | ||
| GenBank mRNA | AY631397.1 | - | 6 | 555 | ||
| Gene Symbol: LOC689629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760982.1 | XP_008759204.1 | 8 | 1235 | ||

LOC499607
Genomic Map