siRNA Details
| Gene Symbol: LOC365791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761013.1 | XP_008759235.1 | 18 | 2636 | ||
| GenBank mRNA | BC081933.1 | - | 2507 | 7114567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC365791| Gene Symbol: LOC365791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761013.1 | XP_008759235.1 | 18 | 2636 | ||
| GenBank mRNA | BC081933.1 | - | 2507 | 7114567 | ||