siRNA Details
Gene Symbol: Lhfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109183.1 | NP_001102653.1 | 2 | 572 | ||
XM_008761002.1 | XP_008759224.1 | 2 | 613 | |||
GenBank mRNA | BC091355.1 | - | 1 | 307 | 7318141 |
