siRNA Details
| Gene Symbol: Tsc22d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191960.1 | NP_001178889.1 | 1 | 1537 | ||
| XM_006232386.2 | XP_006232448.1 | 1 | 2438 | |||
| XM_006232387.2 | XP_006232449.1 | 1 | 2445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsc22d2| Gene Symbol: Tsc22d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191960.1 | NP_001178889.1 | 1 | 1537 | ||
| XM_006232386.2 | XP_006232448.1 | 1 | 2438 | |||
| XM_006232387.2 | XP_006232449.1 | 1 | 2445 | |||