siRNA Details
| Gene Symbol: Wdr49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761118.1 | XP_008759340.1 | 14 | 2206 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr49| Gene Symbol: Wdr49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761118.1 | XP_008759340.1 | 14 | 2206 | ||