siRNA Details
| Gene Symbol: LOC499643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232520.2 | XP_006232582.1 | 4 | 602 | ||
| XM_008761109.1 | XP_008759331.1 | 3 | 221 | |||
| GenBank mRNA | BC085354.1 | - | 4 | 492 | 7132360 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499643| Gene Symbol: LOC499643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232520.2 | XP_006232582.1 | 4 | 602 | ||
| XM_008761109.1 | XP_008759331.1 | 3 | 221 | |||
| GenBank mRNA | BC085354.1 | - | 4 | 492 | 7132360 | |