siRNA Details
| Gene Symbol: RGD1563667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100985.1 | NP_001094455.1 | 2 | 899 | ||
| GenBank mRNA | AY902367.1 | - | 2 | 899 | ||
| FJ004894.1 | - | 774 | ||||
| Gene Symbol: LOC100911650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749346.3 | XP_003749394.2 | 1 | 207 | ||

RGD1563667
Genomic Map