siRNA Details
| Gene Symbol: RGD1563667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100985.1 | NP_001094455.1 | 2 | 487 | ||
| GenBank mRNA | AY902367.1 | - | 2 | 487 | ||
| FJ004894.1 | - | 362 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563667| Gene Symbol: RGD1563667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100985.1 | NP_001094455.1 | 2 | 487 | ||
| GenBank mRNA | AY902367.1 | - | 2 | 487 | ||
| FJ004894.1 | - | 362 | ||||