siRNA Details
| Gene Symbol: Lingo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109189.1 | NP_001102659.1 | 11 | 4466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lingo4| Gene Symbol: Lingo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109189.1 | NP_001102659.1 | 11 | 4466 | ||