siRNA Details
| Gene Symbol: Amy2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031502.1 | NP_113690.1 | 1165 | |||
| GenBank mRNA | FQ233367.1 | - | 8 | 1160 | ||
| J00703.2 | - | 1165 | ||||
| Gene Symbol: LOC103689940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761448.1 | XP_008759670.1 | 8 | 1174 | ||

Amy2a3
Genomic Map