siRNA Details
| Gene Symbol: Synpo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191963.1 | NP_001178892.1 | 4 | 2336 | ||
| XM_008761470.1 | XP_008759692.1 | 5 | 2343 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synpo2| Gene Symbol: Synpo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191963.1 | NP_001178892.1 | 4 | 2336 | ||
| XM_008761470.1 | XP_008759692.1 | 5 | 2343 | |||