siRNA Details
Gene Symbol: Gar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024306.1 | NP_001019477.1 | 4, 5 | 703 | ||
XM_006232006.2 | XP_006232068.1 | 4, 5 | 1431 | |||
XM_008761519.1 | XP_008759741.1 | 4, 5 | 1432 | |||
GenBank mRNA | BC079131.1 | - | 4, 5 | 703 | 7127379 |
