siRNA Details
| Gene Symbol: LOC499715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047950.1 | NP_001041415.1 | 4 | 283 | ||
| GenBank mRNA | AY387081.1 | - | 4 | 283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499715| Gene Symbol: LOC499715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047950.1 | NP_001041415.1 | 4 | 283 | ||
| GenBank mRNA | AY387081.1 | - | 4 | 283 | ||