siRNA Details
| Gene Symbol: Clca4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077356.2 | NP_001070824.1 | 7 | 1080 | ||
| GenBank mRNA | AB256513.1 | - | 7 | 1081 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clca4l| Gene Symbol: Clca4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077356.2 | NP_001070824.1 | 7 | 1080 | ||
| GenBank mRNA | AB256513.1 | - | 7 | 1081 | ||