siRNA Details
| Gene Symbol: RGD1560065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134616.1 | NP_001128088.1 | 2 | 303 | ||
| XM_006233461.2 | XP_006233523.1 | 2 | 382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560065| Gene Symbol: RGD1560065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134616.1 | NP_001128088.1 | 2 | 303 | ||
| XM_006233461.2 | XP_006233523.1 | 2 | 382 | |||