siRNA Details
Gene Symbol: Hsp90aa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175761.2 | NP_786937.1 | 9 | 1689 | ||
GenBank mRNA | AJ428213.1 | - | 9 | 1689 | ||
BC072489.1 | - | 9 | 1671 | 7105514 | ||
BC085120.1 | - | 9 | 1671 | 7114546 | ||
FQ212905.1 | - | 3 | 319 |
Gene Symbol: LOC103692716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764969.1 | XP_008763191.1 | 1741 |
