siRNA Details
| Gene Symbol: LOC499742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024308.1 | NP_001019479.1 | 1, 2 | 131 | ||
| GenBank mRNA | AY539888.1 | - | 1, 2 | 131 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499742| Gene Symbol: LOC499742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024308.1 | NP_001019479.1 | 1, 2 | 131 | ||
| GenBank mRNA | AY539888.1 | - | 1, 2 | 131 | ||