siRNA Details
| Gene Symbol: LOC499746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233649.2 | XP_006233711.1 | 5 | 649 | ||
| XM_006233650.2 | XP_006233712.1 | 5 | 649 | |||
| XM_575081.6 | XP_575081.1 | 5 | 649 | |||
| GenBank mRNA | BC082090.1 | - | 4 | 680 | ||
| BC100078.1 | - | 5 | 600 | |||
| BC160838.1 | - | 1053 | ||||

LOC499746
Genomic Map