siRNA Details
| Gene Symbol: LOC499746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233649.2 | XP_006233711.1 | 5 | 957 | ||
| XM_575081.6 | XP_575081.1 | 6 | 850 | |||
| GenBank mRNA | BC082090.1 | - | 5 | 881 | ||
| BC100078.1 | - | 5 | 908 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499746| Gene Symbol: LOC499746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233649.2 | XP_006233711.1 | 5 | 957 | ||
| XM_575081.6 | XP_575081.1 | 6 | 850 | |||
| GenBank mRNA | BC082090.1 | - | 5 | 881 | ||
| BC100078.1 | - | 5 | 908 | |||