siRNA Details
| Gene Symbol: Tprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024309.2 | NP_001019480.2 | 1 | 1682 | ||
| GenBank mRNA | BC083928.1 | - | 1 | 1016 | 7135551 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tprn| Gene Symbol: Tprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024309.2 | NP_001019480.2 | 1 | 1682 | ||
| GenBank mRNA | BC083928.1 | - | 1 | 1016 | 7135551 | |