siRNA Details
| Gene Symbol: Ccdc183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025040.2 | NP_001020211.2 | 9 | 908 | ||
| XM_006233635.1 | XP_006233697.1 | 10 | 1557 | |||
| GenBank mRNA | BC160897.1 | - | 9 | 908 | 7453119 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc183| Gene Symbol: Ccdc183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025040.2 | NP_001020211.2 | 9 | 908 | ||
| XM_006233635.1 | XP_006233697.1 | 10 | 1557 | |||
| GenBank mRNA | BC160897.1 | - | 9 | 908 | 7453119 | |