siRNA Details
| Gene Symbol: RGD1565512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134617.1 | NP_001128089.1 | 4 | 987 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565512| Gene Symbol: RGD1565512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134617.1 | NP_001128089.1 | 4 | 987 | ||