siRNA Details
| Gene Symbol: Med22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077679.1 | NP_001071147.1 | 1 | 241 | ||
| GenBank mRNA | BC127519.1 | - | 1 | 241 | 7104131 | |
| Gene Symbol: LOC100911917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749430.3 | XP_003749478.1 | 413 | |||

Med22
Genomic Map