siRNA Details
| Gene Symbol: RGD1564114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127563.1 | NP_001121035.1 | 3 | 382 | ||
| GenBank mRNA | BC166521.1 | - | 3 | 382 | 7133997 | |
| Gene Symbol: LOC100909960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749443.3 | XP_003749491.1 | 2 | 156 | ||

RGD1564114
Genomic Map