siRNA Details
| Gene Symbol: Ttf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233882.2 | XP_006233944.1 | 2 | 670 | ||
| XM_006233883.2 | XP_006233945.1 | 2 | 670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttf1| Gene Symbol: Ttf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233882.2 | XP_006233944.1 | 2 | 670 | ||
| XM_006233883.2 | XP_006233945.1 | 2 | 670 | |||