siRNA Details
| Gene Symbol: Garnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761788.1 | XP_008760010.1 | 7, 8 | 850 | ||
| GenBank mRNA | FQ211851.1 | - | 6, 7 | 822 | ||
| FQ211906.1 | - | 4, 5 | 601 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Garnl3| Gene Symbol: Garnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761788.1 | XP_008760010.1 | 7, 8 | 850 | ||
| GenBank mRNA | FQ211851.1 | - | 6, 7 | 822 | ||
| FQ211906.1 | - | 4, 5 | 601 | |||