siRNA Details
| Gene Symbol: Garnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761788.1 | XP_008760010.1 | 17 | 1608 | ||
| GenBank mRNA | FQ211851.1 | - | 16 | 1583 | ||
| FQ211906.1 | - | 14 | 1362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Garnl3| Gene Symbol: Garnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761788.1 | XP_008760010.1 | 17 | 1608 | ||
| GenBank mRNA | FQ211851.1 | - | 16 | 1583 | ||
| FQ211906.1 | - | 14 | 1362 | |||