siRNA Details
| Gene Symbol: RGD1561276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575198.5 | XP_575198.5 | 2 | 677 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561276| Gene Symbol: RGD1561276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575198.5 | XP_575198.5 | 2 | 677 | ||