siRNA Details
| Gene Symbol: RGD1566059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234699.1 | XP_006234761.1 | 2 | 385 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1566059| Gene Symbol: RGD1566059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234699.1 | XP_006234761.1 | 2 | 385 | ||