siRNA Details
Gene Symbol: LOC100911267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762135.1 | XP_008760357.1 | 6 | 688 |
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109204.1 | NP_001102674.1 | 10 | 1151 | ||
XM_006234784.2 | XP_006234846.1 | 11 | 1329 | |||
GenBank mRNA | BC168875.1 | - | 10 | 1115 | 9022295 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
