siRNA Details
| Gene Symbol: Ubr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178072.1 | NP_001171543.1 | 3, 4 | 414 | ||
| GenBank mRNA | AB549211.1 | - | 3, 4 | 414 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubr1| Gene Symbol: Ubr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178072.1 | NP_001171543.1 | 3, 4 | 414 | ||
| GenBank mRNA | AB549211.1 | - | 3, 4 | 414 | ||