siRNA Details
| Gene Symbol: Ap5s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134618.1 | NP_001128090.1 | 2 | 476 | ||
| XM_006235038.2 | XP_006235100.1 | 2 | 495 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap5s1| Gene Symbol: Ap5s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134618.1 | NP_001128090.1 | 2 | 476 | ||
| XM_006235038.2 | XP_006235100.1 | 2 | 495 | |||