siRNA Details
| Gene Symbol: Zfp133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024313.1 | NP_001019484.2 | 4 | 799 | ||
| GenBank mRNA | BC079070.1 | - | 4 | 799 | 7121746 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp133| Gene Symbol: Zfp133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024313.1 | NP_001019484.2 | 4 | 799 | ||
| GenBank mRNA | BC079070.1 | - | 4 | 799 | 7121746 | |