siRNA Details
Gene Symbol: Bpifa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235367.1 | XP_006235429.1 | 7 | 930 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bpifa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235367.1 | XP_006235429.1 | 7 | 930 |